Anti-SYP Rabbit Monoclonal Antibody [clone: YE269]

Lieferant: Abcam
ABCAAB32127-100EA 793 EUR
ABCAAB32127-100
Anti-SYP Rabbit Monoclonal Antibody [clone: YE269]
Antikörper
Type: Primary
Antigen: SYP (synaptophysin)
Clonality: monoclonal
Clone: YE269
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR