Anti-HIST3H3 Rabbit monoclonal antibody unconjugated [clone: Y28]

Lieferant: Abcam
ABCAAB32129-100EA 697 EUR
ABCAAB32129-100
Anti-HIST3H3 Rabbit monoclonal antibody unconjugated [clone: Y28]
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: monoclonal
Clone: Y28
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR