Anti-EIF4EBP1 Rabbit monoclonal antibody unconjugated [clone: Y330]

Lieferant: Abcam
ABCAAB32130-100EA 757 EUR
ABCAAB32130-100
Anti-EIF4EBP1 Rabbit monoclonal antibody unconjugated [clone: Y330]
Antikörper
Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: monoclonal
Clone: Y330
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR