Anti-TP53 Rabbit Monoclonal Antibody [clone: Y179]

Lieferant: Abcam
ABCAAB32132-100EA 902 EUR
ABCAAB32132-100
Anti-TP53 Rabbit Monoclonal Antibody [clone: Y179]
Antikörper
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: Y179
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR