Anti-Rsk 2 / MAPKAP Kinase 1b Rabbit Monoclonal Antibody [clone: Y83]

Lieferant: Abcam
ABCAAB32133-100EA 749 EUR
ABCAAB32133-100
Anti-Rsk 2 / MAPKAP Kinase 1b Rabbit Monoclonal Antibody [clone: Y83]
Antikörper
Type: Primary
Antigen: Rsk 2 / MAPKAP Kinase 1b
Clonality: monoclonal
Clone: Y83
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR