Anti-APP Rabbit monoclonal antibody unconjugated [clone: Y188]

Lieferant: Abcam
ABCAAB32136-100EA 793 EUR
ABCAAB32136-100
Anti-APP Rabbit monoclonal antibody unconjugated [clone: Y188]
Antikörper
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: Y188
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR