Anti-JUN Rabbit Monoclonal Antibody [clone: E254]
Lieferant: Abcam
ABCAAB32137-100EA 981 EUR
ABCAAB32137-100
Anti-JUN Rabbit Monoclonal Antibody [clone: E254]
Antikörper
Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: monoclonal
Clone: E254
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen