Anti-PARP1 Rabbit Monoclonal Antibody [clone: E102]
Lieferant: Abcam
ABCAAB32138-100EA 1030 EUR
ABCAAB32138-100
Anti-PARP1 Rabbit Monoclonal Antibody [clone: E102]
Antikörper
Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: monoclonal
Clone: E102
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen