Anti-DAXX Rabbit monoclonal antibody unconjugated [clone: E94]

Lieferant: Abcam
ABCAAB32140-100EA 917 EUR
ABCAAB32140-100
Anti-DAXX Rabbit monoclonal antibody unconjugated [clone: E94]
Antikörper
Type: Primary
Antigen: DAXX (death-domain associated protein)
Clonality: monoclonal
Clone: E94
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR