Anti-GFP Rabbit monoclonal antibody unconjugated [clone: E385]

Lieferant: Abcam
ABCAAB32146-100EA 749 EUR
ABCAAB32146-100
Anti-GFP Rabbit monoclonal antibody unconjugated [clone: E385]
Antikörper
Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: monoclonal
Clone: E385
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR