Anti-CDK2 Rabbit monoclonal antibody unconjugated [clone: E304]

Lieferant: Abcam
ABCAAB32147-100EA 779 EUR
ABCAAB32147-100
Anti-CDK2 Rabbit monoclonal antibody unconjugated [clone: E304]
Antikörper
Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: monoclonal
Clone: E304
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR