Anti-CHEK2 Rabbit Monoclonal Antibody [clone: Y171]

Lieferant: Abcam
ABCAAB32148-100EA 901 EUR
ABCAAB32148-100
Anti-CHEK2 Rabbit Monoclonal Antibody [clone: Y171]
Antikörper
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: monoclonal
Clone: Y171
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR