Anti-pro Caspase 3 Rabbit Monoclonal Antibody [clone: E61]

Lieferant: Abcam
ABCAAB32150-100EA 779 EUR
ABCAAB32150-100
Anti-pro Caspase 3 Rabbit Monoclonal Antibody [clone: E61]
Antikörper
Type: Primary
Antigen: pro Caspase 3
Clonality: monoclonal
Clone: E61
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR