Anti-FLT1 Rabbit monoclonal antibody unconjugated [clone: Y103]
Lieferant: Abcam
ABCAAB32152-100EA 1040 EUR
ABCAAB32152-100
Anti-FLT1 Rabbit monoclonal antibody unconjugated [clone: Y103]
Antikörper
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: monoclonal
Clone: Y103
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen