Anti-pro Caspase 10 Rabbit Monoclonal Antibody [clone: E35]

Lieferant: Abcam
ABCAAB32155-100EA 819 EUR
ABCAAB32155-100
Anti-pro Caspase 10 Rabbit Monoclonal Antibody [clone: E35]
Antikörper
Type: Primary
Antigen: pro Caspase 10
Clonality: monoclonal
Clone: E35
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR