Anti-EIF2S1 Rabbit monoclonal antibody unconjugated [clone: E90]
Lieferant: Abcam
ABCAAB32157-100EA 1050 EUR
ABCAAB32157-100
Anti-EIF2S1 Rabbit monoclonal antibody unconjugated [clone: E90]
Antikörper
Type: Primary
Antigen: EIF2S1 (eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa)
Clonality: monoclonal
Clone: E90
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen