Anti-BCL2L11 Rabbit monoclonal antibody unconjugated [clone: Y36]

Lieferant: Abcam
ABCAAB32158-100EA 779 EUR
ABCAAB32158-100
Anti-BCL2L11 Rabbit monoclonal antibody unconjugated [clone: Y36]
Antikörper
Type: Primary
Antigen: BCL2L11 (BCL2-like 11 (apoptosis facilitator))
Clonality: monoclonal
Clone: Y36
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR