Anti-PTPN11 Rabbit Monoclonal Antibody [clone: Y477]
Lieferant: Abcam
ABCAAB32159-100EA 958 EUR
ABCAAB32159-100
Anti-PTPN11 Rabbit Monoclonal Antibody [clone: Y477]
Antikörper
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: monoclonal
Clone: Y477
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen