Anti-AXIN2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32197-100EA 703 EUR
ABCAAB32197-100
Anti-AXIN2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AXIN2 (axin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR