Anti-EGFR Rabbit monoclonal antibody unconjugated [clone: E234]

Lieferant: Abcam
ABCAAB32198-100EA 688 EUR
ABCAAB32198-100
Anti-EGFR Rabbit monoclonal antibody unconjugated [clone: E234]
Antikörper
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: E234
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR