Anti-PTEN Rabbit Monoclonal Antibody [clone: Y184]

Lieferant: Abcam
ABCAAB32199-100EA 808 EUR
ABCAAB32199-100
Anti-PTEN Rabbit Monoclonal Antibody [clone: Y184]
Antikörper
Type: Primary
Antigen: PTEN (phosphatase and tensin homolog)
Clonality: monoclonal
Clone: Y184
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR