Anti-RPS6KA1 Rabbit Monoclonal Antibody [clone: E239]
Lieferant: Abcam
ABCAAB32203-100EA 909 EUR
ABCAAB32203-100
Anti-RPS6KA1 Rabbit Monoclonal Antibody [clone: E239]
Antikörper
Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: monoclonal
Clone: E239
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen