Anti-IDE Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32216-100EA 1020 EUR
ABCAAB32216-100
Anti-IDE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IDE (insulin-degrading enzyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR