Anti-ARHGAP1 Rabbit Monoclonal Antibody [clone: EP489Y]
Lieferant: Abcam
ABCAAB32328-100EA 918 EUR
ABCAAB32328-100
Anti-ARHGAP1 Rabbit Monoclonal Antibody [clone: EP489Y]
Antikörper
Type: Primary
Antigen: ARHGAP1 (Rho GTPase activating protein 1)
Clonality: monoclonal
Clone: EP489Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen