Anti-Caldesmon Rabbit Monoclonal Antibody [clone: E89]

Lieferant: Abcam
ABCAAB32330-100EA 703 EUR
ABCAAB32330-100
Anti-Caldesmon Rabbit Monoclonal Antibody [clone: E89]
Antikörper
Type: Primary
Antigen: Caldesmon
Clonality: monoclonal
Clone: E89
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR