Anti-GOLGA2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB32337-100EA 657 EUR
ABCAAB32337-100
Anti-GOLGA2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GOLGA2 (golgin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR