Anti-STX16 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32340-100EA 682 EUR
ABCAAB32340-100
Anti-STX16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STX16 (syntaxin 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR