Anti-CASP3 Rabbit monoclonal antibody unconjugated [clone: E87]

Lieferant: Abcam
ABCAAB32351-100EA 1020 EUR
ABCAAB32351-100
Anti-CASP3 Rabbit monoclonal antibody unconjugated [clone: E87]
Antikörper
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: E87
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR