Anti-HIV1 p24 Rabbit monoclonal antibody unconjugated [clone: P131]

Lieferant: Abcam
ABCAAB32352-100EA 873 EUR
ABCAAB32352-100
Anti-HIV1 p24 Rabbit monoclonal antibody unconjugated [clone: P131]
Antikörper
Type: Primary
Antigen: HIV1 p24
Clonality: monoclonal
Clone: P131
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR