Anti-CKAP4 Rabbit monoclonal antibody unconjugated [clone: Y289]

Lieferant: Abcam
ABCAAB32353-100EA 688 EUR
ABCAAB32353-100
Anti-CKAP4 Rabbit monoclonal antibody unconjugated [clone: Y289]
Antikörper
Type: Primary
Antigen: CKAP4 (cytoskeleton-associated protein 4)
Clonality: monoclonal
Clone: Y289
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR