Anti-HIST3H3 Rabbit monoclonal antibody unconjugated [clone: Y47]
Lieferant: Abcam
ABCAAB32356-100EA 997 EUR
ABCAAB32356-100
Anti-HIST3H3 Rabbit monoclonal antibody unconjugated [clone: Y47]
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: monoclonal
Clone: Y47
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen