Anti-CEBPB Rabbit monoclonal antibody unconjugated [clone: E299]

Lieferant: Abcam
ABCAAB32358-100EA 763 EUR
ABCAAB32358-100
Anti-CEBPB Rabbit monoclonal antibody unconjugated [clone: E299]
Antikörper
Type: Primary
Antigen: CEBPB (CCAAT/enhancer binding protein (C/EBP), beta)
Clonality: monoclonal
Clone: E299
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR