Anti-NFKB1 Rabbit Monoclonal Antibody [clone: E381]

Lieferant: Abcam
ABCAAB32360-100EA 773 EUR
ABCAAB32360-100
Anti-NFKB1 Rabbit Monoclonal Antibody [clone: E381]
Antikörper
Type: Primary
Antigen: NFKB1 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 1)
Clonality: monoclonal
Clone: E381
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR