Anti-DES Rabbit monoclonal antibody unconjugated [clone: Y266]

Lieferant: Abcam
ABCAAB32362-100EA 793 EUR
ABCAAB32362-100
Anti-DES Rabbit monoclonal antibody unconjugated [clone: Y266]
Antikörper
Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone: Y266
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR