Anti-GAB2 Rabbit monoclonal antibody unconjugated [clone: EP369Y]

Lieferant: Abcam
ABCAAB32365-100EA 626 EUR
ABCAAB32365-100
Anti-GAB2 Rabbit monoclonal antibody unconjugated [clone: EP369Y]
Antikörper
Type: Primary
Antigen: GAB2 (GRB2-associated binding protein 2)
Clonality: monoclonal
Clone: EP369Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR