Anti-CASP6 Rabbit monoclonal antibody unconjugated [clone: E180]

Lieferant: Abcam
ABCAAB32366-100EA 688 EUR
ABCAAB32366-100
Anti-CASP6 Rabbit monoclonal antibody unconjugated [clone: E180]
Antikörper
Type: Primary
Antigen: CASP6 (caspase 6, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: E180
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR