Anti-STAT2 Rabbit Monoclonal Antibody [clone: Y141]

Lieferant: Abcam
ABCAAB32367-100EA 672 EUR
ABCAAB32367-100
Anti-STAT2 Rabbit Monoclonal Antibody [clone: Y141]
Antikörper
Type: Primary
Antigen: STAT2 (signal transducer and activator of transcription 2, 113kDa)
Clonality: monoclonal
Clone: Y141
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR