Anti-HDAC3 Rabbit monoclonal antibody unconjugated [clone: Y415]

Lieferant: Abcam
ABCAAB32369-100EA 763 EUR
ABCAAB32369-100
Anti-HDAC3 Rabbit monoclonal antibody unconjugated [clone: Y415]
Antikörper
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: monoclonal
Clone: Y415
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR