Anti-BCL2L1 Rabbit monoclonal antibody unconjugated [clone: E18]

Lieferant: Abcam
ABCAAB32370-100EA 749 EUR
ABCAAB32370-100
Anti-BCL2L1 Rabbit monoclonal antibody unconjugated [clone: E18]
Antikörper
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: monoclonal
Clone: E18
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR