Anti-SGK1 Rabbit Monoclonal Antibody [clone: Y238]

Lieferant: Abcam
ABCAAB32374-100EA 933 EUR
ABCAAB32374-100
Anti-SGK1 Rabbit Monoclonal Antibody [clone: Y238]
Antikörper
Type: Primary
Antigen: SGK1 (serum/glucocorticoid regulated kinase 1)
Clonality: monoclonal
Clone: Y238
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR