Anti-ERBB4 Rabbit Monoclonal Antibody [clone: E200]

Lieferant: Abcam
ABCAAB32375-100EA 732 EUR
ABCAAB32375-100
Anti-ERBB4 Rabbit Monoclonal Antibody [clone: E200]
Antikörper
Type: Primary
Antigen: ERBB4 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4)
Clonality: monoclonal
Clone: E200
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR