Anti-PRKCA Rabbit Monoclonal Antibody [clone: Y124]
Lieferant: Abcam
ABCAAB32376-100EA 1070 EUR
ABCAAB32376-100
Anti-PRKCA Rabbit Monoclonal Antibody [clone: Y124]
Antikörper
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: monoclonal
Clone: Y124
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen