Anti-14-3-3 alpha/beta Rabbit Monoclonal Antibody [clone: E58]

Lieferant: Abcam
ABCAAB32377-100EA 850 EUR
ABCAAB32377-100
Anti-14-3-3 alpha/beta Rabbit Monoclonal Antibody [clone: E58]
Antikörper
Type: Primary
Antigen: 14-3-3 alpha/beta (14-3-3 alpha/beta)
Clonality: monoclonal
Clone: E58
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR