Anti-CDK1 Rabbit monoclonal antibody unconjugated [clone: E161]

Lieferant: Abcam
ABCAAB32384-100EA 912 EUR
ABCAAB32384-100
Anti-CDK1 Rabbit monoclonal antibody unconjugated [clone: E161]
Antikörper
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: monoclonal
Clone: E161
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR