Anti-JUN Rabbit Monoclonal Antibody [clone: Y172]

Lieferant: Abcam
ABCAAB32385-100EA 749 EUR
ABCAAB32385-100
Anti-JUN Rabbit Monoclonal Antibody [clone: Y172]
Antikörper
Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: monoclonal
Clone: Y172
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR