Anti-GSK3B Rabbit Monoclonal Antibody [clone: Y174]
Lieferant: Abcam
ABCAAB32391-100EA 1010 EUR
ABCAAB32391-100
Anti-GSK3B Rabbit Monoclonal Antibody [clone: Y174]
Antikörper
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: monoclonal
Clone: Y174
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen