Anti-EPCAM Rabbit Monoclonal Antibody [clone: E144]
Lieferant: Abcam
ABCAAB32392-100EA 981 EUR
ABCAAB32392-100
Anti-EPCAM Rabbit Monoclonal Antibody [clone: E144]
Antikörper
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone: E144
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen