Anti-IRAKM Rabbit Monoclonal Antibody [clone: Y278]

Lieferant: Abcam
ABCAAB32394-100EA 732 EUR
ABCAAB32394-100
Anti-IRAKM Rabbit Monoclonal Antibody [clone: Y278]
Antikörper
Type: Primary
Antigen: IRAKM
Clonality: monoclonal
Clone: Y278
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR