Anti-PKN2 Rabbit Monoclonal Antibody [clone: E352]

Lieferant: Abcam
ABCAAB32395-100EA 688 EUR
ABCAAB32395-100
Anti-PKN2 Rabbit Monoclonal Antibody [clone: E352]
Antikörper
Type: Primary
Antigen: PKN2 (protein kinase N2)
Clonality: monoclonal
Clone: E352
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR