Anti-ESR1 Rabbit Monoclonal Antibody [clone: E91]

Lieferant: Abcam
ABCAAB32396-100EA 697 EUR
ABCAAB32396-100
Anti-ESR1 Rabbit Monoclonal Antibody [clone: E91]
Antikörper
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: monoclonal
Clone: E91
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR